Aluminum n-butoxide, 90+%
Aluminum n-butoxide, 90+%
Aluminum n-butoxide, 90+%
Thermo Scientific Chemicals

Aluminum n-butoxide, 90+%

CAS: 3085-30-1 | C12H30AlO3 | 249.351 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 087754.18
also known as 087754-18
Price (USD)
481.65
Special offer
Online exclusive
Ends: 30-Jun-2026
567.00
Save 85.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
481.65
Special offer
Online exclusive
Ends: 30-Jun-2026
567.00
Save 85.35 (15%)
Each
Chemical Identifiers
CAS3085-30-1
IUPAC Namealuminium(3+) tris(butan-1-olate)
Molecular FormulaC12H27AlO3
InChI KeyMYWQGROTKMBNKN-UHFFFAOYSA-N
SMILES[Al+3].CCCC[O-].CCCC[O-].CCCC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
FormFused solid
Assay from Supplier's CofA≥90%
Melting Point102?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only