Ammonium diamminetetrathiocyanatochromate(III) monohydrate, ACS, 93.0% min
Thermo Scientific Chemicals

Ammonium diamminetetrathiocyanatochromate(III) monohydrate, ACS, 93.0% min

Used to precipitate primary and secondary amines as their ammonium salts | CAS: 13573-17-6 | C4H10CrN7S4·H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 087777.14
also known as 087777-14
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
77.50
Save 11.85 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
77.50
Save 11.85 (15%)
Each
Chemical Identifiers
CAS13573-17-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)93.0% C₄H₁₀CrN₇S₄.H₂O min
CommentSensitivity P.T.
Impurity contentInsoluble in dilute hydrochloric acid ≤ 0.05%
Ammonium diamminetetrathiocyanatochromate(III) monohydrate is used to precipitate primary and secondary amines as their ammonium salts. It is a color reagent and precipitant for mercury. It is also used as a precipitant for primary and secondary amines and certain amino acids. It is a reagent used for quantitative studies, including titrimetric, gravimetric, and spechtometric assays. Additionally, Reinecke salt has also been shown to determine the concentration of Dimenhydrinate using a spectrophotometric procedure. Other studies use Reinecke salt as a chemical actinometer to calculate the photodegradation of Manidipine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only