Decaborane, 98%, Thermo Scientific Chemicals
Decaborane, 98%, Thermo Scientific Chemicals
Decaborane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Decaborane, 98%, Thermo Scientific Chemicals

In rocket propellants, and as a catalyst in olefin polymerization
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 087892.14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS17702-41-9
Molecular FormulaB10H14
Molecular Weight (g/mol)122.22
MDL NumberMFCD00011314
SpecificationsSpecification SheetSpecification Sheet
Formcrystalline powder and chunks
Appearance (Color)White to pale yellow
Assay (unspecified)> 97.5%
Decaborane is used as a fuel for aneutronic fusion. It is also used in the preparation of vinyl decaborane as well as in the preparation of semiconductors that have low energy ion implantation. It is widely used in rocket propellants and serves as a catalyst in olefin polymerization reactions. It is a reactant for the preparation of carborane-conjugated quinoline carboxamide ligands of translocator protein for boron neutron capture therapy (BNCT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only