Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III)
Thermo Scientific Chemicals

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III)

CAS: 15522-71-1 | C33H60EuO6 | 704.801 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 087898.03
also known as 087898-03
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
71.20
Save 10.55 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
71.20
Save 10.55 (15%)
Each
Add to cart
Specifications
CAS15522-71-1
Chemical Name or MaterialTris (2,2,6,6-tetramethyl-3,5-heptanedionato)europium (III)
EINECS Number239-565-5
Formula Weight701.78
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only