Bis(cyclopentadienyl)nickel, dry
Bis(cyclopentadienyl)nickel, dry
Bis(cyclopentadienyl)nickel, dry
Thermo Scientific Chemicals

Bis(cyclopentadienyl)nickel, dry

CAS: 1271-28-9 | C10H10Ni | 188.883 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
50 g
Catalog number 087899.18
also known as 087899-18
Price (USD)
349.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
349.00
Each
Chemical Identifiers
CAS1271-28-9
IUPAC Namenickel(2+) bis(cyclopenta-2,4-dien-1-ide)
Molecular FormulaC10H10Ni
InChI KeyKZPXREABEBSAQM-UHFFFAOYSA-N
SMILES[Ni++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals
Appearance (Color)Green to brown
Elemental Analysis> 98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only