Perchloric acid, ACS, 69.0-72.0%, redistilled
Perchloric acid, ACS, 69.0-72.0%, redistilled
Perchloric acid, ACS, 69.0-72.0%, redistilled
Thermo Scientific Chemicals

Perchloric acid, ACS, 69.0-72.0%, redistilled

CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Catalog number 087963.AP
also known as 087963-AP
Price (USD)
164.00
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
164.00
Each
Chemical Identifiers
CAS7601-90-3
IUPAC Nameperchloric acid
Molecular FormulaClHO4
InChI KeyVLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILESO[Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)10
Assay (unspecified)69.0-72.0% HClO₄
Impurity contentCl 0.001%
Impurity contentFe 1ppm
Impurity contentNitrogen compounds (as N) 0.001%
View more
Perchloric acid is used as a precursor for ammonium perchlorate which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals and alkali metals such as sodium and potassium. It is also used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only