Boron, AAS solution standard, Specpure™ B 1000μg/mL
Thermo Scientific Chemicals

Boron, AAS solution standard, Specpure™ B 1000μg/mL

H3BO3 in H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 088055.AP
also known as 088055-AP
Price (USD)
78.65
Online Exclusive
87.60
Save 8.95 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
78.65
Online Exclusive
87.60
Save 8.95 (10%)
Each
Specifications
Chemical Name or MaterialBoron, AAS standard solution
Health Hazard 1H360FD
Health Hazard 3P201-P202-P281-P308+P313-P501c
MDL NumberMFCD00134034
Molecular FormulaH3BO3 in H2O
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only