Cobalt, AAS standard solution, Specpure™ Co 1000μg/mL
Cobalt, AAS standard solution, Specpure™ Co 1000μg/mL
Cobalt, AAS standard solution, Specpure™ Co 1000μg/mL
Thermo Scientific Chemicals

Cobalt, AAS standard solution, Specpure™ Co 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 088060.AP
also known as 088060-AP
Price (USD)
85.65
Online Exclusive
95.30
Save 9.65 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
85.65
Online Exclusive
95.30
Save 9.65 (10%)
Each
Specifications
Chemical Name or MaterialCobalt, AAS standard solution
DOT InformationHazard Class: 8; Packaging Group: II
Health Hazard 1H290-H314-H335-H350i-H360F
Health Hazard 2GHS H Statement
H334-H314-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P234-P260-P264b-P271-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only