Copper, AAS standard solution, Specpure™ Cu 1000μg/mL
Copper, AAS standard solution, Specpure™ Cu 1000μg/mL
Copper, AAS standard solution, Specpure™ Cu 1000μg/mL
Thermo Scientific Chemicals

Copper, AAS standard solution, Specpure™ Cu 1000μg/mL

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 088061.AE
also known as 088061-AE
Price (USD)
48.20
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
48.20
Each
Specifications
CAS7440-50-8
CAS7732-18-5
ChEBICHEBI:30052
Chemical Name or MaterialCopper
ColorColorless
View more
Copper, AAS standard solution is used to determine copper in samples of water and water-suspendedsediment with a specific conductance not greater than 10,000 μS/cm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only