Gold, AAS standard solution, Specpure™ Au 1000μg/mL
Gold, AAS standard solution, Specpure™ Au 1000μg/mL
Gold, AAS standard solution, Specpure™ Au 1000μg/mL
Thermo Scientific Chemicals

Gold, AAS standard solution, Specpure™ Au 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 088068.AE
also known as 088068-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialGold, AAS standard solution
DOT InformationHazard Class: 8; Packaging Group: II
EINECS Number231-595-7
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.
View more
Gold, AAS standard solution is used for the determination of gold by flame atomic absorption spectrometry (FAAS) and graphite furnace atomic absorption spectrometry (GFAAS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only