Lead, AAS standard solution, Specpure™ Pb 1000μg/mL
Lead, AAS standard solution, Specpure™ Pb 1000μg/mL
Lead, AAS standard solution, Specpure™ Pb 1000μg/mL
Thermo Scientific Chemicals

Lead, AAS standard solution, Specpure™ Pb 1000μg/mL

CAS: 7439-92-1 | Pb | 207.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 088075.AE
also known as 088075-AE
Price (USD)
46.50
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
46.50
Each
Specifications
CAS7439-92-1
CAS7697-37-2
CAS7732-18-5
ChEBICHEBI:27889
Chemical Name or MaterialLead
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only