Lithium, AAS standard solution, Specpure™ Li 1000μg/mL
Lithium, AAS standard solution, Specpure™ Li 1000μg/mL
Lithium, AAS standard solution, Specpure™ Li 1000μg/mL
Thermo Scientific Chemicals

Lithium, AAS standard solution, Specpure™ Li 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 088076.AP
also known as 088076-AP
Price (USD)
82.60
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
82.60
Each
Specifications
Chemical Name or MaterialLithium, AAS standard solution
DOT InformationHazard Class: 8
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only