Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL
Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL
Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL
Thermo Scientific Chemicals

Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL

CAS: 7439-96-5 | Mn | 54.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 088078.AE
also known as 088078-AE
Price (USD)
45.60
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
45.60
Each
Specifications
CAS7439-96-5
CAS7697-37-2
CAS7732-18-5
ChEBICHEBI:35154
Chemical Name or MaterialManganese
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only