Mercury, AAS standard solution, Specpure™ Hg 1000μg/mL
Thermo Scientific Chemicals

Mercury, AAS standard solution, Specpure™ Hg 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 088079.AE
also known as 088079-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialMercury, AAS standard solution
DOT InformationHazard Class: 8; Packaging Group: II
EINECS Number231-714-2
Health Hazard 1H290-H314-H335-H373
Health Hazard 2GHS H Statement
H360-H314
May damage fertility or the unborn child.
Causes severe skin burns and eye damage.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only