Nickel, AAS standard solution, Specpure™ Ni 1000μg/mL
Nickel, AAS standard solution, Specpure™ Ni 1000μg/mL
Nickel, AAS standard solution, Specpure™ Ni 1000μg/mL
Thermo Scientific Chemicals

Nickel, AAS standard solution, Specpure™ Ni 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 088082.AE
also known as 088082-AE
Price (USD)
45.60
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
45.60
Each
Specifications
Chemical Name or MaterialNickel, AAS standard solution
ColorColorless
DOT InformationHazard Class: 8; Packaging Group: II
Health Hazard 1H290-H314-H335-H350i-H360D
Health Hazard 2GHS H Statement
H314-H317-H208
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only