Niobium, AAS standard solution, Specpure™ Nb 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium, AAS standard solution, Specpure™ Nb 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 088083.AE
also known as 088083-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialNiobium, AAS standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROFLUORIC ACID
Health Hazard 1H290-H311-H315-H319-H332
Health Hazard 2GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P234-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P361-P363-P390-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only