Vanadium, AAS standard solution, Specpure™ V 1000μg/mL
Thermo Scientific Chemicals

Vanadium, AAS standard solution, Specpure™ V 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 088116.AP
also known as 088116-AP
Price (USD)
73.65
Special offer
Online exclusive
Ends: 30-Jun-2026
86.10
Save 12.45 (14%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
73.65
Special offer
Online exclusive
Ends: 30-Jun-2026
86.10
Save 12.45 (14%)
Each
Specifications
Chemical Name or MaterialVanadium, AAS standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H361
Causes severe skin burns and eye damage.
Suspected of damaging fertility or the unborn child.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only