Yttrium, AAS standard solution, Specpure™ Y 1000μg/mL
Yttrium, AAS standard solution, Specpure™ Y 1000μg/mL
Yttrium, AAS standard solution, Specpure™ Y 1000μg/mL
Thermo Scientific Chemicals

Yttrium, AAS standard solution, Specpure™ Y 1000μg/mL

CAS: 7697-37-2 | HNO3 | 63.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 088117.AP
also known as 088117-AP
Price (USD)
97.50
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
97.50
Each
Specifications
Assay Percent RangeY 1000;mug/mL
CAS7697-37-2
ChEBICHEBI:48107
Chemical Name or MaterialYttrium
DOT InformationDOT Class: 8, Packaging Group: III
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only