B·R·A·H·M·S™ GM KRYPTOR™ QC
Thermo Scientific™

B·R·A·H·M·S™ GM KRYPTOR™ QC

B·R·A·H·M·S provides a complete portfolio of biomarkers for the most reliable results in prenatal screening. Get a reliable trisomy screening with the multiparametric control B·R·A·H·M·S GM KRYPTOR QC containing PAPP-A, Free βhCG and AFP.
Have Questions?
Catalog NumberForm
088192NLyophilized
Catalog number 088192N
Price (MXN)
-
Form:
Lyophilized

B·R·A·H·M·S provides a complete portfolio of biomarkers for the most reliable results in prenatal screening. Get a reliable trisomy screening with the multiparametric control B·R·A·H·M·S GM KRYPTOR QC containing PAPP-A, Free βhCG and AFP to be used with B·R·A·H·M·S PAPP-A KRYPTOR, B·R·A·H·M·S Free βhCG KRYPTOR and B·R·A·H·M·S AFP KRYPTOR.

US Only: Product is not 510k cleared in US.

  • Save cost and time with a multiparametric control containing 3 parameters: PAPP-A, Free βhCG and AFP
  • Covering the clinical ranges of interest with 3 levels of concentration
  • Long shelf life with lyophilized controls
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
Specifications
DescriptionControls for B·R·A·H·M·S AFP KRYPTOR (3 levels), B·R·A·H·M·S Free ΒhCG KRYPTOR (3 levels), B·R·A·H·M·S PAPP-A KRYPTOR (3 levels)
For Use With (Application)Quality Control for B·R·A·H·M·S AFP KRYPTOR assay, B·R·A·H·M·S Free ΒhCG KRYPTOR assay, B·R·A·H·M·S PAPP-A KRYPTOR assay
Detectable Analytesalpha-fetoprotein, beta-hCG (Free), PAPP-A
FormLyophilized
Unit SizeBox with 6 vials
Contents & Storage
Controls kit, 6 vials, 2°C to 8°C