Iron nitride
Iron nitride
Iron nitride
Thermo Scientific Chemicals

Iron nitride

CAS: 37245-77-5 | Fe6H2N2-2 | 365.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 088198.22
also known as 088198-22
Price (USD)
690.65
Special offer
Online exclusive
Ends: 30-Jun-2026
812.00
Save 121.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
690.65
Special offer
Online exclusive
Ends: 30-Jun-2026
812.00
Save 121.35 (15%)
Each
Chemical Identifiers
CAS37245-77-5
SpecificationsSpecification SheetSpecification Sheet
Impurity content0.1% max.
Iron nitride is used in the synthesis of ammonia and carbon nanofibers. It is involved in the creation of ferrofluids, a liquid rocket fuel. It finds applications in making the strongest naturally magnetic material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only