Indium(III) selenide, 99.99% (metals basis)
Indium(III) selenide, 99.99% (metals basis)
Indium(III) selenide, 99.99% (metals basis)
Thermo Scientific Chemicals

Indium(III) selenide, 99.99% (metals basis)

CAS: 12056-07-4 | In2Se3
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 088280.06
also known as 088280-06
Price (USD)
123.65
Special offer
Online exclusive
Ends: 28-Dec-2026
145.00
Save 21.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
123.65
Special offer
Online exclusive
Ends: 28-Dec-2026
145.00
Save 21.35
Each
Chemical Identifiers
CAS12056-07-4
IUPAC Namediindium(3+) triselandiide
Molecular FormulaIn2Se3
InChI KeyAKUCEXGLFUSJCD-UHFFFAOYSA-N
SMILES[Se--].[Se--].[Se--].[In+3].[In+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Indium(III) selenide is used in photovoltaic devices. It is also used for optoelectronics and in solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only