Lead(II) fluoride, 99% min
Lead(II) fluoride, 99% min
Lead(II) fluoride, 99% min
Thermo Scientific Chemicals

Lead(II) fluoride, 99% min

CAS: 7783-46-2 | F2Pb | 245.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 088282.A1
also known as 088282-A1
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS7783-46-2
IUPAC Nameλ²-lead(2+) difluoride
Molecular FormulaF2Pb
InChI KeyFPHIOHCCQGUGKU-UHFFFAOYSA-L
SMILES[F-].[F-].[Pb++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99% min
Lead(II) fluoride is a specialty product for proteomics research. It is used in low melting glasses, in glass coatings to reflect infrared rays, in phosphors for television-tube screens and as a catalyst for the manufacture of picoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only