Zinc sulfide, 99.9% (metals basis)
Zinc sulfide, 99.9% (metals basis)
Zinc sulfide, 99.9% (metals basis)
Thermo Scientific Chemicals

Zinc sulfide, 99.9% (metals basis)

CAS: 1314-98-3 | SZn | 97.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 088299.30
also known as 088299-30
Price (USD)
181.65
Online Exclusive
202.00
Save 20.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
181.65
Online Exclusive
202.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS1314-98-3
IUPAC Namezinc(2+) sulfanediide
Molecular FormulaSZn
InChI KeyDRDVZXDWVBGGMH-UHFFFAOYSA-N
SMILES[S--].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Zinc sulfide is used in luminescent materials, infra red optical materials, semiconductors, as a pigment, and as a window for visible optics and infrared optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only