Silicon(IV) oxide, 99% (metals basis)
Silicon(IV) oxide, 99% (metals basis)
Silicon(IV) oxide, 99% (metals basis)
Thermo Scientific Chemicals

Silicon(IV) oxide, 99% (metals basis)

CAS: 14808-60-7 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 088316.30
also known as 088316-30
Price (USD)
79.20
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
79.20
Each
Chemical Identifiers
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤1.0%
Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only