Aluminum Silicon powder, -325 mesh, 99% (metals basis)
Aluminum Silicon powder, -325 mesh, 99% (metals basis)
Aluminum Silicon powder, -325 mesh, 99% (metals basis)
Thermo Scientific Chemicals

Aluminum Silicon powder, -325 mesh, 99% (metals basis)

CAS: 11145-27-0 | AlSi | 55.067 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 088322.22
also known as 088322-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS11145-27-0
IUPAC Namealuminium silicon
Molecular FormulaAlSi
InChI KeyCSDREXVUYHZDNP-UHFFFAOYSA-N
SMILES[Al].[Si]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities≤1.0%
Appearance (Color)Grey
Elemental AnalysisSi: 11.0-13.0%
Aluminum Silicon powder is a reducing agent which is used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only