Bronze powder, -100 mesh, 99.0% (metals basis)
Thermo Scientific Chemicals

Bronze powder, -100 mesh, 99.0% (metals basis)

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 088359.30
also known as 088359-30
Price (USD)
67.65
Online exclusive
75.50
Save 7.85
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
67.65
Online exclusive
75.50
Save 7.85
Each
Specifications
AppearanceBronze colored powder
Assay Percent Range99% (Metals basis)
Chemical Name or MaterialBronze
Health Hazard 1H228-H315-H319-H335
Health Hazard 3P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
Bronze powder suitable for a range of applications including resin-casting, decorative coatings and powder metallurgy. Also used for hydraulic seals, surface engineering, decorative, welding and brazing, diamond tooling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only