Titanium aluminide, -325 mesh, 99.5% (metals basis)
Titanium aluminide, -325 mesh, 99.5% (metals basis)
Titanium aluminide, -325 mesh, 99.5% (metals basis)
Thermo Scientific Chemicals

Titanium aluminide, -325 mesh, 99.5% (metals basis)

CAS: 12003-96-2 | AlTi3+3 | 170.583 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 088393.18
also known as 088393-18
Price (USD)
243.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
243.00
Each
Chemical Identifiers
CAS12003-96-2
SpecificationsSpecification SheetSpecification Sheet
Particle Sizeca. 325 Mesh
FormPowder
Appearance (Color)Silver grey
Assay from Supplier's CofA≥99.5%
It finds use in several applications including automobiles and aircraft. Also used in air frames, investment casting, turbocharger rotors, and wire/fiber metal matrix composites

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only