MAbPac™ HIC-10 HPLC Column
Thermo Scientific™

MAbPac™ HIC-10 HPLC Column

Green features

Separate monoclonal antibodies (mAbs) and related biologics with high resolution, excellent biocompatibility, and selectivity using the unique chemistry of this hydrophobic interaction column.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
088480100 mm4.6 mm
088481250 mm4.6 mm
SP6997100 mm7.8 mm
088482
also known as 088482   
10 mm4.6 mm
Catalog number 088480
Price (MXN)
-
Length (Metric):
100 mm
Diameter (Metric):
4.6 mm

Designed for separation of monoclonal antibodies (mAbs) and related biologics by hydrophobic interaction, the Thermo Scientific™ MAbPac™ HIC-10 LC column is a high resolution silica column. The MAbPac HIC-10 column's unique chemistry provides high resolution, excellent biocompatibility, and selectivity that is different from other HIC columns on the market.

Unique Chemistry Designed for Optimal Selectivity of mAbs

The MAbPac HIC-10 column is based on high-purity, spherical, wide-pore (1,000Å), 5μm silica particles functionalized with proprietary alkyl amide groups. Its advanced surface bonding technology leads to excellent chemical stability, high efficiency, and unique selectivity optimized for mAbs and different from other HIC phases.

  • Excellent biocompatibility and extremely low carryover
  • High resolution and high efficiency
  • Compatible with both organic solvent and aqueous mobile phases
  • Rugged column stability
Specifications
Column FormatAnalytical Column
Column TypeHydrophobic Interaction
Green FeaturesFSC, ACT Ecolabel (Gold)
Max. Pressure6000 psi (415 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
QuantityEach
Stationary PhaseSilica
Temperature60°C
Diameter (Metric)4.6 mm
Length (Metric)100 mm
Particle Size5 μm
PhaseHydrophobic Interaction
Pore Size1000 Å
Product LineMAbPac
TechniqueBio-LC
pH2 to 8
Unit SizeEach