Tris(trifluoromethylhydroxymethylene-d-camphorato)europium(III)
Tris(trifluoromethylhydroxymethylene-d-camphorato)europium(III)
Tris(trifluoromethylhydroxymethylene-d-camphorato)europium(III)
Thermo Scientific Chemicals

Tris(trifluoromethylhydroxymethylene-d-camphorato)europium(III)

CAS: 34830-11-0 | C36H45EuF9O6 | 896.7 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
Catalog number 088481.03
also known as 088481-03
Price (USD)
145.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
145.00
Each
Specifications
CAS34830-11-0
Chemical Name or MaterialTris (trifluoromethylhydroxymethylene-d-camphorato)europium (III)
EINECS Number252-232-9
Formula Weight893.68
IUPAC Nameeuropium;(2E)-4,7,7-trimethyl-2-(2,2,2-trifluoro-1-hydroxyethylidene)bicyclo[2.2.1]heptan-3-one
View more
Optically active NMR shift reagent. Used with Ag(fod) for the enantiomeric resolution of sec-butylisothiouronium chloride. Also reactant involved in synthesis of europium acetyl camphorate phosphine oxide complexes and reactions with 1,10-N,N-phenanthroline dioxide derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only