Dionex™ IonPac™ AS22-Fast-4μm IC Columns
Actual product may vary
Thermo Scientific™

Dionex™ IonPac™ AS22-Fast-4μm IC Columns

Green features

Obtain fast, high-resolution separation of common inorganic anions in diverse matrices using this medium capacity anion-exchange column with a carbonate/bicarbonate eluent.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
08848730 mm4 mm
088486150 mm4 mm
088488150 mm2 mm
08848930 mm2 mm
Catalog number 088487
Price (USD)
682.00
Each
Length (Metric):
30 mm
Diameter (Metric):
4 mm
Price (USD)
682.00
Each

Attain the optimal combination of chromatographic speed and resolution using the Thermo Scientific™ Dionex™ IonPac™ AS22-Fast-4μm column. Designed specifically for compliance monitoring of inorganic anions in accordance with U.S. EPA Method 300.0 (A), this moderate capacity column combines the selectivity of the Dionex IonPac AS22-Fast column with fast high resolution separations. Compared to the Dionex IonPac AS22-Fast column, the new Dionex IonPac AS22-Fast-4μm column uses smaller resin particles for more efficient separations resulting in faster run times, more accurate peak integration and more reliable results.

Fast separation of common inorganic anions with excellent resolution
  • Obtain the highest peak efficiency, through the use of smaller resin particles, of common inorganic anions in diverse sample matrices
  • Attain fast isocratic separation of the common inorganic anions in less than 5 min
  • Perform compliance monitoring of common anions in drinking water as specified in U.S. EPA Method 300.0 (A)
  • Achieve excellent separation of inorganic anions and low-molecular weight acids in diverse sample matrices
  • Obtain high peak efficiency and increased peak resolution for more consistent integration and more reliable results
  • Transfer application easily--this 4μm column has the the same selectivity as the Dionex IonPac AS22-Fast column
  • Achieve excellent resolution of the carbonate peak away from the common inorganic anions
  • Reduce run time without loss of resolution compared to the Dionex IonPac AS22-Fast column
  • Faster alternative, using carbonate/bicarbonate eluents, for Dionex IonPac AS4A-SC, AS12A, AS14, AS14A and AS22 inorganic anion applications
  • Compatible with organic solvents to enhance analyte solubility, modify column selectivity, or allow effective column cleanup
Use with a high-pressure IC system

Higher resolution is achieved through the use of smaller resin particles, which can produce a column backpressure over 3000 psi. When operating the Dionex IonPac AS22-Fast-4μm column, a high-pressure IC system, such as the Thermo Scientific™ Dionex™ ICS-6000 HPIC system is required.

Use the format that fits your lab
  • 4 x 150 mm, 2 x 150 mm analytical columns
  • 4 x 30 mm, 2 x 30 mm guard columns
  • Shorter 150 mm columns facilitate faster runs while the smaller particle size maintains a higher resolution separation

Note: For simple, accurate eluent preparation, use the Dionex IonPac AS22 Eluent Concentrate or the Dionex Eluent Generator (Dionex EGC 500 K2CO3 cartridge with Dionex EPM 500 pH modifier).

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceU.S. EPA Method 300.0 (A)
Column ChemistryCarbonate/Bicarbonate
Column ConstructionPEEK
Flow Rate2.0 mL/min.
For Use With (Application)Anion Exchange
For Use With (Equipment)IonPac AS22-Fast-4μm Analytical Column
Green FeaturesFSC
Max. Pressure5000 psi (345 bar)
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
Solvents0 to 100% of HPLC Solvents
Column TypeGuard Column
Diameter (Metric)4 mm
FormatStandard Bore
Length (Metric)30 mm
pH0 to 14
Unit SizeEach