Dionex™ IonPac™ CS16-4μm and CS16-Fast-4μm IC Columns
Actual product may vary
Thermo Scientific™

Dionex™ IonPac™ CS16-4μm and CS16-Fast-4μm IC Columns

Green features
Analyze disparate concentration ratios of close-eluting cations such as sodium and ammonium with these very high-capacity cation exchange columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column CapacityFlow Rate
08860230 mm2 mm160 μeq0.16 to 0.25 mL/min.
088582250 mm2 mm1340 μeq0.16 to 0.25 mL/min.
08858350 mm2 mm270 μeq0.16 to 0.25 mL/min.
088584250 mm4 mm5370 μeq0.64 to 1.0 mL/min.
08858550 mm4 mm1070 μeq0.64 to 1.0 mL/min.
088599150 mm4 mm3220 μeq0.64 to 1.0 mL/min.
08860030 mm4 mm650 μeq0.64 to 1.0 mL/min.
088601150 mm2 mm800 μeq0.16 to 0.25 mL/min.
088615250 mm0.4 mm50 μeq0.006 to 0.010 mL/min.
08861650 mm0.4 mm10 μeq0.006 to 0.010 mL/min.
Catalog number 088602
Price (USD)
-
Length (Metric):
30 mm
Diameter (Metric):
2 mm
Column Capacity:
160 μeq
Flow Rate:
0.16 to 0.25 mL/min.

Analyze disparate concentration ratios up to 10,000:1 of close-eluting cations such as ammonium and sodium using Thermo Scientific™ Dionex™ IonPac™ CS16-4μm and CS16-Fast-4μm columns. Designed specifically for the analysis of cations at diverse concentration ratios, this high-capacity column combines the selectivity of the Dionex IonPac CS16 column with fast, high resolution separations. Compared to the Dionex IonPac CS16 column, the new Dionex IonPac CS16-4μm and CS16-Fast-4μm columns use smaller resin particles for more efficient separations resulting in faster run times, more accurate peak integration, and more reliable results.

Highest Resolution Separation of Ammonium and Sodium at Disparate Concentration Ratios up to 10,000:1 in Diverse Matrices
  • Recommended for trace-level ammonium in high concentrations of sodium without sample pretreatment
  • Recommended for trace-level sodium in high concentrations of ammonium without sample pretreatment
  • Use the 250 mm Dionex IonPac CS16-4μm for complex matrices
  • Use the 150 mm Dionex IonPac CS16-Fast-4μm for faster analyses in simple matrices.
  • Easily transfer applications from the Dionex IonPac CS16 column to the 4 μm columns due to the similar selectivity
  • High capacity facilitates injection of more concentrated samples without column overloading or peak broadening
  • Compatible with organic solvents, excluding alcohols, to enhance analyte solubility, modify column selectivity, and facilitate effective column clean-up
Use with a High-Pressure IC System

Higher resolution is achieved through the use of smaller resin particles, which produce a column backpressure over 3000 psi at the standard flow rates. A high-pressure IC system, such as the Thermo Scientific™ Dionex™ ICS-6000+ HPIC™ system, is required to operate this column under standard conditions and take advantage of elevated flow rates and faster run times.

Reduce Operating Costs With Lower Eluent Requirements

The Dionex IonPac CS16-4μm capillary columns provide the same performance as the standard bore and microbore Dionex IonPac CS16-4μm columns because they are packed with the same material. Operating costs are reduced because the capillary system operates at only 1% of the eluent flow rate of the standard bore system, requiring less eluent and producing less waste.

Multiple Formats Available for Application Flexibility
  • Dionex IonPac CS16-4μm: Available in 4 mm, 2 mm, and 0.4 mm formats
  • Dionex IonPac CS16-Fast-4μm: Available in 4 mm and 2 mm formats
  • Longer 250 mm columns facilitate higher resolution separations of more complex sample matrices
  • Shorter 150 mm columns provide faster analyses of simple matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity160 μeq
Column ChemistryAcidic
Column ConstructionPEEK
Crosslinking55% DVB
Flow Rate0.16 to 0.25 mL/min.
For Use With (Application)Cation Exchange
For Use With (Equipment)Dionex IonPac CS16-Fast-4μm Microbore Separator Column
Green FeaturesFSC
HydrophobicityMedium
Ion Exchange GroupCarboxylic Acid
Max. Pressure5000 psi (345 bar)
Packing MaterialEthylvinylbenzene/Divinylbenzene
Particle Size4 μm
QuantityEach 1
Solvent CompatibilityAcidic Eluents (pH 0 to 7), 0 to 100% Acetonitrile, 0 to 100% Acetone, Alcohols Should be Avoided
Stationary PhaseIonPac CG16-Fast-4μm
Column TypeGuard Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)30 mm
Unit SizeEach 1