Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N - FAQs
Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N - FAQs
View additional product information for Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N - FAQs (088602.LS, 088602.LQ)
2 product FAQs found
What are the instructions for dilution of 1 unit of Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N (Cat. No. 088602.LS, 088602.LQ)?
The following is the dilution protocol:
- Transfer entire contents of 1 unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask.
- Repeat rinse three times. Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 0.5N Potassium hydroxide.
What is 1 unit of Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N (Cat. No. 088602.LS, 088602.LQ)?
1 unit of Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N, when diluted to 1 L, will yield a final concentration of 0.5N Potassium hydroxide solution.