Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N - FAQs
Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N - FAQs
View additional product information for Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N, Thermo Scientific Chemicals - FAQs (088620.LS)
2 product FAQs found
What is the protocol for diluting one unit of Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N (Cat. No. 088620.LS, 088620-LS)?
The following is the dilution protocol:
- Transfer entire contents of one unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask.
- Repeat rinse three times. Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 0.5N Sodium hydroxide.
What is 1 unit of Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N?
1 unit of Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N when diluted to 1 L will yield a final concentration of 0.5N Sodium hydroxide solution.