Sodium oxalate, Acculute Standard Volumetric Solution, Final Concentration 0.1N
Sodium oxalate, Acculute Standard Volumetric Solution, Final Concentration 0.1N
Sodium oxalate, Acculute Standard Volumetric Solution, Final Concentration 0.1N
Thermo Scientific Chemicals

Sodium oxalate, Acculute Standard Volumetric Solution, Final Concentration 0.1N

CAS: 62-76-0 | C2Na2O4 | 134.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
6 Units
1 Unit
Catalog number 088626.LS
also known as 088626-LS
Price (USD)
352.65
Online Exclusive
392.00
Save 39.35 (10%)
Each
Quantity:
6 Units
Request bulk or custom format
Price (USD)
352.65
Online Exclusive
392.00
Save 39.35 (10%)
Each
Chemical Identifiers
CAS62-76-0
IUPAC Namedisodium oxalate
Molecular FormulaC2Na2O4
InChI KeyZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA0.1000N ± 0.0002N (after dilution)
FormLiquid
Appearance (Color)Colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Frequently asked questions (FAQs)

What are the instructions for dilution of 1 unit of Sodium oxalate, Acculute Standard Volumetric Solution, Final Concentration 0.1N (Cat. No. 088626.LS, 088626.LQ)?

The following is the dilution protocol:
- Transfer entire contents of 1 unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask.
- Repeat rinse three times. Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 0.1N Sodium oxalate.

What is 1 unit of Sodium oxalate, Acculute Standard Volumetric Solution, Final Concentration 0.1N (088626.LS, 088626.LQ)?

1 unit of Sodium oxalate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, when diluted to 1 L, will yield a final concentration of 0.1N Sodium oxalate solution.