Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N
Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N
Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N
Thermo Scientific Chemicals

Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N

CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 U
6 U
Catalog number 088628.LQ
also known as 088628-LQ
Price (USD)
69.30
Each
Quantity:
1 U
Request bulk or custom format
Price (USD)
69.30
Each
Chemical Identifiers
CAS7772-98-7
IUPAC Namedisodium sulfanidesulfonate
Molecular FormulaNa2O3S2
InChI KeyAKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([S-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA0.1000N ? 0.0002N (after dilution)
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What are the instructions for dilution of 1 unit of Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N (Cat. No. 088628.LS, 088628.LQ)?

The following is the dilution protocol:
- Transfer entire contents of 1 unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask.
- Repeat rinse three times. Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 0.1N Sodium thiosulfate.

What is 1 unit of Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N (Cat. No. 088628.LS, 088628.LQ)?

1 unit of Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, when diluted to 1 L, will yield a final concentration of 0.1N Sodium thiosulfate solution.