MAbPac™ Reversed Phase HPLC Columns
Thermo Scientific™

MAbPac™ Reversed Phase HPLC Columns

Green features

Use the new Thermo Scientific™ MAbPac™ RP columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Stationary Phase
088644100 mm3 mmPhenyl (Ph)
08864550 mm3 mmPhenyl (Ph)
08864610 mm3 mmPhenyl (Ph)
088647100 mm2.1 mmPhenyl (Ph)
08864850 mm2.1 mmPhenyl (Ph)
08864910 mm2.1 mmPhenyl (Ph)
30318250 mm1 mmPSDVB (Polystyrene Divinylbenzene)
303183100 mm1 mmPSDVB (Polystyrene Divinylbenzene)
303184150 mm1 mmPSDVB (Polystyrene Divinylbenzene)
303269150 mm3 mm
303270150 mm2.1 mm
Catalog number 088644
Price (MXN)
-
Length (Metric):
100 mm
Diameter (Metric):
3 mm
Stationary Phase:
Phenyl (Ph)

Use the new Thermo Scientific™ MAbPac™ Reversed Phase columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over. The surface chemistry, particle size, and pore size, have been optimized for high resolution separations.

Compatible with organic solvent used in HPLC and MS under RP conditions

  • Resin is based on a hydrophobic supermacroporous 4μm polymer particles
  • Stable at extreme pH (0 to 14) and high temperature (up to 110 °C)

Efficient with low carry over

  • 1,500Å large pore size polymeric resin
  • Efficient separation of protein molecules with very low carry over

Designed for reversed-phase biomolecule LC and LC/MS separations

  • Superior resolution power for monoclonal antibodies and related substances
  • High efficiency with low carry-over
  • Excellent MS compatibility
  • Wide operating pH range: 0 to 14
  • High temperature stability: up to 110°C
  • High throughput
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeReversed Phase
Green FeaturesFSC
Max. Pressure4000 psi (275 bar)
Packing MaterialDVB (Divinylbenzene) Polymer
QuantityEach
Stationary PhasePhenyl (Ph)
Temperature<110°C
Diameter (Metric)3 mm
Length (Metric)100 mm
Particle Size4 μm
PhaseReversed Phase
Pore Size1500 Å
Product LineMAbPac
TechniqueBio-LC
pH0 to 14
Unit SizeEach