Dionex™ IonPac™ AS28-Fast-4μm IC Columns
Actual product may vary
Thermo Scientific™

Dionex™ IonPac™ AS28-Fast-4μm IC Columns

Green features

Get fast, high efficiency, high resolution from these 4μm particle columns for trace analysis of common inorganic anions and low molecular weight organic acids.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
08875235 mm0.4 mm
088751150 mm0.4 mm
088749150 mm2 mm
08875030 mm2 mm
088747150 mm4 mm
08874830 mm4 mm
Catalog number 088752
Price (USD)
-
Length (Metric):
35 mm
Diameter (Metric):
0.4 mm

Analyze trace inorganic anions and low molecular weight organic acids using the Thermo Scientific™ Dionex™ IonPac™ AS28-Fast-4μm ion chromatography (IC) columns. Designed specifically for trace level analysis of high purity water from semiconductor and power industries, this column combines the selectivity of the Thermo Scientific™ Dionex™ IonPac™ AS15 column, with fast, high efficiency, and high resolution separations. The Dionex IonPac AS28-Fast-4μm columns uses smaller 4μm resin particles for more efficient separations resulting in more accurate peak integration and more reliable results.

Applications for Dionex IonPac AS28-Fast-4μm IC Columns
  • Separation of common anions and low molecular weight organic acids
  • For both high purity water matrices and complex matrices including chemical and petrochemical samples
  • Compatible with organic solvents to enhance analyte solubility, modify column selectivity, and facilitate effective column clean-up

Superior Performance
  • Improved efficiencies and resolution compared to the Dionex IonPac AS15 column
  • Easily transfer applications from the Dionex IonPac AS15 column to the AS28-Fast-4μm column due to the similar selectivity

For Use With a High-Pressure IC System

Higher resolution is achieved through the use of smaller resin particles, which produce a column backpressure over 3000 psi at the standard flow rates. A high-pressure IC system, such as the Thermo Scientific™ Dionex™ ICS-5000+ HPIC™ or the Thermo Scientific Dionex ICS-4000 Capillary HPIC system is required to operate this column under standard conditions and take advantage of elevated flow rates and faster run times.


Reduce Operating Costs With Lower Eluent Requirements

The Dionex IonPac AS28-Fast-4μm capillary columns provide the same performance as the standard bore and microbore Dionex IonPac AS28-Fast-4μm columns because they are packed with the same material. Operating costs are reduced because the capillary system operates at only 1% of the eluent flow rate of the standard bore system, requiring less eluent and producing less waste.


Multiple Formats Available
  • 4 x 150mm and 2 x 150mm analytical columns
  • 4 x 30mm and 2 x 30mm analytical guard columns
  • 0.4 x 150mm capillary columns
  • 0.4 x 35mm capillary guard columns
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation, and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Anion Exchange Capacity0.2 μeq
Column Capacity0.2 μeq
Column ChemistryHydroxide
Column ConstructionPEEK
Crosslinking55% DVB
Flow Rate0.012 to 0.02 mL/min.
For Use With (Application)Anion Exchange
For Use With (Equipment)Dionex IonPac AS28-Fast-4μm Capillary Column
Green FeaturesFSC
HydrophobicityMedium-High
Ion Exchange GroupAlkanol Quaternary Ammonium Ion
Max. Pressure5000 psi (345 bar)
Packing MaterialEthylvinylbenzene/Divinylbenzene
Particle Size4 μm
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% Acetonitrile, Methanol, and Isopropanol
Stationary PhaseMicroporous Ethylvinylbenzene Polymer Cross-Linked with Divinylbenzene
Column TypeGuard Column
Diameter (Metric)0.4 mm
FormatCapillary
Length (Metric)35 mm
Unit SizeEach 1