Lithium trifluoroacetate monohydrate, 97%
Lithium trifluoroacetate monohydrate, 97%
Lithium trifluoroacetate monohydrate, 97%
Thermo Scientific Chemicals

Lithium trifluoroacetate monohydrate, 97%

CAS: 2923-17-3 | C2F3LiO2 | 119.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 088938.09
also known as 088938-09
Price (USD)
45.60
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
45.60
Each
Specifications
CAS2923-17-3
Chemical Name or MaterialLithium trifluoroacetate monohydrate
ColorWhite
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECS Number220-878-0
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only