Thermo Scientific Chemicals

Strontium chromate, 95%, Thermo Scientific Chemicals

Catalog number: 089026.30
250 g, Each
Thermo Scientific Chemicals

Strontium chromate, 95%, Thermo Scientific Chemicals

Catalog number: 089026.30
250 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 089026.30
also known as 089026-30
Price (USD)
Price: 72.70
Online price: 62.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7789-06-2
IUPAC Name
strontium(2+) dioxochromiumbis(olate)
Molecular Formula
CrO4Sr
InChI Key
NVKTUNLPFJHLCG-UHFFFAOYSA-N
SMILES
[Sr++].[O-][Cr]([O-])(=O)=O
Assay (unspecified)
95.0% min

Description

Strontium chromate is used an primer for zinc, magnesium, aluminum, and alloys used in aircraft manufacture. Used as a colorant in polyvinyl chloride resins. It is also used in aluminum flake coatings. paints, varnishes and oil colors. It is a used in water based wash primers, metal conditioners, additive to control the sulfate content of solutions in electrochemical processes. It is also used in paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text