Sodium trimetaphosphate
Sodium trimetaphosphate
Sodium trimetaphosphate
Thermo Scientific Chemicals

Sodium trimetaphosphate

CAS: 7785-84-4 | Na3O9P3 | 305.882 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 089063.A7
also known as 089063-A7
Price (USD)
298.65
Online Exclusive
332.00
Save 33.35 (10%)
Each
Quantity:
5 kg
Request bulk or custom format
Price (USD)
298.65
Online Exclusive
332.00
Save 33.35 (10%)
Each
Chemical Identifiers
CAS7785-84-4
IUPAC Nametrisodium trioxo-1,3,5,2λ⁵,4λ⁵,6λ⁵-trioxatriphosphinane-2,4,6-tris(olate)
Molecular FormulaNa3O9P3
InChI KeyUGTZMIPZNRIWHX-UHFFFAOYSA-K
SMILES[Na+].[Na+].[Na+].[O-]P1(=O)OP([O-])(=O)OP([O-])(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)P₂O₅ 68.0-70.0
Sodium trimetaphosphate is used as a crosslinking agent in the cross-linked amylase-resistant starch. It acts as an intermediate in food industries. Further, it serves as a corrosion inhibitors, anti-scaling agents, fillers, finishing agents, plating agents and surface treating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only