Aluminum bromide, 99.997% (metals basis)
Aluminum bromide, 99.997% (metals basis)
Aluminum bromide, 99.997% (metals basis)
Thermo Scientific Chemicals

Aluminum bromide, 99.997% (metals basis)

CAS: 7727-15-3 | AlBr3 | 266.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 089065.06
also known as 089065-06
Price (USD)
55.65
Online exclusive
61.20
Save 5.55
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
55.65
Online exclusive
61.20
Save 5.55
Each
Chemical Identifiers
CAS7727-15-3
IUPAC Namealuminium(3+) tribromide
Molecular FormulaAlBr3
InChI KeyPQLAYKMGZDUDLQ-UHFFFAOYSA-K
SMILES[Al+3].[Br-].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.003% max.
Aluminum Bromide, is used as a catalyst for the Friedel-Crafts alkylation reaction, and also for bromination and isomerization in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only