Borane-dimethylamine complex, 97%
Borane-dimethylamine complex, 97%
Borane-dimethylamine complex, 97%
Thermo Scientific Chemicals

Borane-dimethylamine complex, 97%

CAS: 74-94-2 | C2H10BN | 58.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 089149.14
also known as 089149-14
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.70
Save 13.05 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.70
Save 13.05 (15%)
Each
Chemical Identifiers
CAS74-94-2
IUPAC Namedimethylamine borane
Molecular FormulaC2H10BN
InChI KeyFTDUHBOCJSQEKS-UHFFFAOYSA-N
SMILESB.CNC
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥96.0%
Borane-dimethylamine complex is used in a nickel-catalyzed reduction of electron-deficient aryl tosylates. It also serves as a reducing agent in electrolytic deposition of metals, alloys, semiconductors and insulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only