Titanium(IV) isobutoxide
Titanium(IV) isobutoxide
Titanium(IV) isobutoxide
Thermo Scientific Chemicals

Titanium(IV) isobutoxide

CAS: 7425-80-1 | C16H36O4Si | 320.545 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 089455.A1
also known as 089455-A1
Price (USD)
359.65
Special offer
Online exclusive
Ends: 30-Jun-2026
423.00
Save 63.35 (15%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
359.65
Special offer
Online exclusive
Ends: 30-Jun-2026
423.00
Save 63.35 (15%)
Each
Chemical Identifiers
CAS7425-80-1
IUPAC Nametitanium(4+) tetrakis(2-methylpropan-1-olate)
Molecular FormulaC16H36O4Ti
InChI KeyQUVMSYUGOKEMPX-UHFFFAOYSA-N
SMILES[Ti+4].CC(C)C[O-].CC(C)C[O-].CC(C)C[O-].CC(C)C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid to low melting solid
Infrared spectrumConforms
Appearance (Color)Pale yellow
Titanium(IV) isobutoxide general application in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only