Bismuth(III) chloride, anhydrous, 99.999% (metals basis)
Bismuth(III) chloride, anhydrous, 99.999% (metals basis)
Bismuth(III) chloride, anhydrous, 99.999% (metals basis)
Thermo Scientific Chemicals

Bismuth(III) chloride, anhydrous, 99.999% (metals basis)

CAS: 7787-60-2 | BiCl3 | 315.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 089485.06
also known as 089485-06
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
76.40
Save 11.75
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
76.40
Save 11.75
Each
Chemical Identifiers
CAS7787-60-2
IUPAC Namebismuth(3+) trichloride
Molecular FormulaBiCl3
InChI KeyJHXKRIRFYBPWGE-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
It is used as a catalyst in organic chemistry. It is a good source of bismuth and is used to make other bismuth compounds. Also used as a catalyst and to make other bismuth salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only