Sodium hydroxide, 5.0 N in aqueous solution, WFI, Biotech Reagent, GMP, J.T.Baker™
Avantor™

Sodium hydroxide, 5.0 N in aqueous solution, WFI, Biotech Reagent, GMP, J.T.Baker™

Made from Water for Injection U.S.P. and Sodium Hydroxide pellets, N.F. (Multi-Compendial). Meets BP, Ph.Eur. and JP Chemical specifications.
Have Questions?
變更視圖buttonViewtableView
Quantity:
19 L
Packaging:
Poly pail (Jerrican)
Poly pail (Jerrican), case
產品號碼 0895-07
價格 (HKD)
-
Quantity:
19 L
Packaging:
Poly pail (Jerrican)
Made from Water for Injection U.S.P. and Sodium Hydroxide pellets, N.F. (Multi-Compendial). Meets BP, Ph.Eur. and JP Chemical specifications.
Made from Water for Injection U.S.P. and Sodium Hydroxide pellets, N.F. (Multi-Compendial). Meets BP, Ph.Eur. and JP Chemical specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
規格
Boiling Point109°C (1013 hPa)
CAS1310-73-2
Chemical Name or MaterialSodium hydroxide
ColorColorless
Molecular FormulaNaOH
PackagingPoly pail (Jerrican)
SynonymSodium hydrate, Soda lye, Caustic soda, Lye
GradeBiotech
Molecular Weight (g/mol)40
Quantity19 L
Unit SizeEach