Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangeAu:Pt:Pd; 40:40:20 wt%
Chemical Name or MaterialGold Platinum Palladium powder
DOT InformationTransport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
Health Hazard 1H228
View more