Lanthanum(III) sulfate, anhydrous, 99.9% (REO)
Lanthanum(III) sulfate, anhydrous, 99.9% (REO)
Lanthanum(III) sulfate, anhydrous, 99.9% (REO)
Thermo Scientific Chemicals

Lanthanum(III) sulfate, anhydrous, 99.9% (REO)

CAS: 10099-60-2 | La2O12S3 | 565.979 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 089760.18
also known as 089760-18
Price (USD)
109.65
Online exclusive
122.00
Save 12.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
109.65
Online exclusive
122.00
Save 12.35
Each
Chemical Identifiers
CAS10099-60-2
IUPAC Namedilanthanum(3+) trisulfate
Molecular FormulaLa2O12S3
InChI KeyVQEHIYWBGOJJDM-UHFFFAOYSA-H
SMILES[La+3].[La+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Used for atomic weight determinations for lanthanum. Also used as catalyst and in electronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only