Vanadium(V) triisopropoxide oxide, 96%
Vanadium(V) triisopropoxide oxide, 96%
Vanadium(V) triisopropoxide oxide, 96%
Thermo Scientific Chemicals

Vanadium(V) triisopropoxide oxide, 96%

CAS: 5588-84-1 | C9H24O4V | 247.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
500 g
Catalog number 089798.14
also known as 089798-14
Price (USD)
244.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
244.00
Each
Chemical Identifiers
CAS5588-84-1
Molecular FormulaC9H24O4V
InChI KeyJOUSPCDMLWUHSO-UHFFFAOYSA-N
SMILESO=[V].CC(C)O.CC(C)O.CC(C)O
Molecular Weight (g/mol)247.23
SpecificationsSpecification SheetSpecification Sheet
Assay by NMR≥95.0% (Vanadium-51 NMR, non-U.S. specification)
Assay from Suppliers CofA≥95.0% (U.S. specification)
CommentMay darken on storage
CommentSpecification differs for U.S. and non-U.S. material where indicated
Proton NMRConforms to structure (non-U.S. specification)
View more
Vanadium(V) triisopropoxide oxide serves as a building block for a series of neutral oxovanadium complexes containing both ethoxide and acetylacetonate ligands. It is also used in catalysis. Further, it acts as a precursor and used in thermal atomic layer deposition of vanadium pentoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only