Barnstead™ Point of Use Water Purification Systems Cleaning Solution
Thermo Scientific™
Barnstead™ Point of Use Water Purification Systems Cleaning Solution
Maintain the quality of your water purification system with Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Cleaning Solution.
Have Questions?
Catalog NumberFor Use With (Equipment)
09.2202LabTower EDI, Pacific TII, Smart2Pure, Pacific RO
Catalog number 09.2202
Price (USD)
-
For Use With (Equipment):
LabTower EDI, Pacific TII, Smart2Pure, Pacific RO
Maintain the quality of your water purification system with Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Cleaning Solution.
This 12-piece cleaning solution is used with the following Barnstead water purification systems:
  • GenPure™
  • GenPure™ Pro
  • GenPure™ xCAD
  • LabTower™ EDI
  • LabTower™ RO
  • LabTower™ TII
  • MicroPure™
  • Pacific™ RO
  • Pacific™ TII
  • Smart2Pure™
Specifications
DescriptionCleaning solution (Europe and Asia Pacific)
For Use With (Equipment)LabTower EDI, Pacific TII, Smart2Pure, Pacific RO
Unit SizeEach