DRI™Serum Toxicology Benzodiazepine Assay
Thermo Scientific™

DRI™Serum Toxicology Benzodiazepine Assay

Utilize this serum toxicology Benzodiazepine assay in emergency room situations where urine samples are not conveniently available.
Have Questions?
Catalog NumberQuantity
092025 x 8 mL
Catalog number 0920
Price (USD)
-
Quantity:
25 x 8 mL
Utilize Thermo Scientific™ DRI™ Serum Toxicology Benzodiazepine Assay in emergency room situations where urine samples are not conveniently available.
  • Serum-based assay
  • Used to indentify class of drug of overdose
  • Conveniently packaged with separate calibrators for ease of use
  • Liquid, ready-to-use and has the flexibility to run on a variety of clinical chemistry analyzers

Ordering Alerts:

0920: Reagent volume; 25, 8mL

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Reagents are ready-to-use; no additional preparation is required.
Specifications
DescriptionSerum Toxicology Benzodiazepine
Quantity25 x 8 mL
Sample TypeSerum
TypeReagent
Detectable AnalytesBenzodiazepines (BZO)
Unit SizeEach
Contents & Storage
2°C to 8°C